Product Details

SNP ID
rs146367075
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:231370634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAAAAACAGCAGTCTATCAAAT[C/T]TGGGTTCAATGTCAGCAAACTGGGC
Phenotype
MIM: 606425
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EGLN1 PubMed Links

Gene Details

Gene
EGLN1
Gene Name
egl-9 family hypoxia inducible factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022051.2 1514 Missense Mutation AAA,AGA K359R NP_071334.1
XM_005273166.4 1514 Missense Mutation AAA,AGA K359R XP_005273223.1
XM_005273167.4 1514 Intron XP_005273224.1

View Full Product Details