Product Details

SNP ID
rs146392390
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:153975887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCTCTACATATCCTGTGGGAC[C/T]ACACTCAGGGGTAGTTTTCTGCCAG
Phenotype
MIM: 607138 MIM: 604671 MIM: 602672 MIM: 604740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
There are no transcripts associated with this gene.

Gene
JTB
Gene Name
jumping translocation breakpoint
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006694.3 946 Missense Mutation AGT,GGT S75G NP_006685.1
Gene
RAB13
Gene Name
RAB13, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
There are no transcripts associated with this gene.

View Full Product Details