Product Details

SNP ID
rs146396685
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:162500042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAACTCCTGGATGTCAGTGTTT[C/T]GGAATTGCTCTTATATTCCAGTCAC
Phenotype
MIM: 608849
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UHMK1 PubMed Links

Gene Details

Gene
UHMK1
Gene Name
U2AF homology motif (UHM) kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184763.1 552 Missense Mutation TCG,TTG S45L NP_001171692.1
NM_144624.2 552 Missense Mutation TCG,TTG S119L NP_653225.2
NM_175866.4 552 Missense Mutation TCG,TTG S119L NP_787062.1

View Full Product Details