Product Details

SNP ID
rs146444571
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:156293114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAACCAAGCCGCCCCCTAGCAGC[A/G]TGAGCCCTGGGGCACCCAGGGCCAC
Phenotype
MIM: 610962 MIM: 615531
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLMP PubMed Links

Gene Details

Gene
GLMP
Gene Name
glycosylated lysosomal membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256604.1 854 Missense Mutation ACG,ATG T335M NP_001243533.1
NM_001256605.1 854 Missense Mutation ACG,ATG T298M NP_001243534.1
NM_001256608.1 854 Silent Mutation CAC,CAT H298H NP_001243537.1
NM_001256609.1 854 Missense Mutation ACG,ATG T303M NP_001243538.1
NM_144580.2 854 Missense Mutation ACG,ATG T384M NP_653181.1
XM_011509117.2 854 Missense Mutation ACG,ATG T207M XP_011507419.1
XM_017000149.1 854 UTR 3 XP_016855638.1
Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

Gene
TMEM79
Gene Name
transmembrane protein 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032323.2 854 Intron NP_115699.1
Gene
VHLL
Gene Name
von Hippel-Lindau tumor suppressor like
There are no transcripts associated with this gene.

View Full Product Details