Product Details

SNP ID
rs146456778
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:10451604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTCCTGCAAATAAAACCTCAC[C/T]CATGAATGCTCACGCAAGTGTAATG
Phenotype
MIM: 609130 MIM: 602784 MIM: 601882
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APITD1 PubMed Links

Gene Details

Gene
APITD1
Gene Name
apoptosis-inducing, TAF9-like domain 1
There are no transcripts associated with this gene.

Gene
APITD1-CORT
Gene Name
APITD1-CORT readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243768.1 832 UTR 3 NP_001230697.1
NM_001270517.1 832 UTR 3 NP_001257446.1
NM_198544.3 832 UTR 3 NP_940946.1
NM_199006.2 832 UTR 3 NP_950171.2
Gene
CORT
Gene Name
cortistatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302.4 832 UTR 3 NP_001293.3
Gene
DFFA
Gene Name
DNA fragmentation factor subunit alpha
There are no transcripts associated with this gene.

View Full Product Details