Product Details

SNP ID
rs146509537
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:46607788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCCCTCTGTTTCATAAAAATA[A/C]ATTTTCCAACCCTCCCCCTATCCCA
Phenotype
MIM: 606724
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MKNK1 PubMed Links

Gene Details

Gene
MKNK1
Gene Name
MAP kinase interacting serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135553.2 5680 Intron NP_001129025.1
NM_003684.5 5680 Intron NP_003675.2
NM_198973.3 5680 Intron NP_945324.1
XM_006711000.1 5680 Intron XP_006711063.1
XM_006711001.2 5680 Intron XP_006711064.1
XM_006711002.2 5680 Intron XP_006711065.1
XM_006711003.1 5680 Intron XP_006711066.1
XM_006711006.3 5680 Intron XP_006711069.1
XM_006711007.1 5680 Intron XP_006711070.1
XM_006711009.3 5680 Intron XP_006711072.1
XM_006711013.1 5680 Intron XP_006711076.1
XM_006711014.1 5680 Intron XP_006711077.1
XM_006711015.1 5680 Intron XP_006711078.1
XM_017002652.1 5680 Intron XP_016858141.1
XM_017002653.1 5680 Intron XP_016858142.1
XM_017002654.1 5680 Intron XP_016858143.1
XM_017002655.1 5680 Intron XP_016858144.1
XM_017002656.1 5680 Intron XP_016858145.1
XM_017002657.1 5680 Intron XP_016858146.1
XM_017002658.1 5680 Intron XP_016858147.1
XM_017002659.1 5680 Intron XP_016858148.1
XM_017002660.1 5680 Intron XP_016858149.1
XM_017002661.1 5680 Intron XP_016858150.1
XM_017002662.1 5680 Intron XP_016858151.1
XM_017002663.1 5680 Intron XP_016858152.1
XM_017002664.1 5680 Intron XP_016858153.1
XM_017002665.1 5680 Intron XP_016858154.1
XM_017002666.1 5680 Intron XP_016858155.1
XM_017002667.1 5680 Intron XP_016858156.1
XM_017002668.1 5680 Intron XP_016858157.1
Gene
MOB3C
Gene Name
MOB kinase activator 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145279.4 5680 UTR 3 NP_660322.2
NM_201403.2 5680 UTR 3 NP_958805.1
XM_017000407.1 5680 UTR 3 XP_016855896.1

View Full Product Details