Product Details

SNP ID
rs146509597
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:248038745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTATCAACTCTTGTGCTCACACA[A/G]TATATGCACTCTGTATCCCATATTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2L13 PubMed Links

Gene Details

Gene
OR2L13
Gene Name
olfactory receptor family 2 subfamily L member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304535.1 704 Intron NP_001291464.1
NM_175911.3 704 Intron NP_787107.1
XM_011544169.2 704 Intron XP_011542471.1
Gene
OR2L2
Gene Name
olfactory receptor family 2 subfamily L member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004686.2 704 Missense Mutation ATA,GTA I160V NP_001004686.1
XM_011544158.2 704 Missense Mutation ATA,GTA I160V XP_011542460.1

View Full Product Details