Product Details

SNP ID
rs146560631
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:36341971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGAGCACTCCTCCGTCACCTT[C/T]GCCTTTGAGGCGGGGAGGGTGGGAA
Phenotype
MIM: 609437
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STK40 PubMed Links

Gene Details

Gene
STK40
Gene Name
serine/threonine kinase 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282546.1 1542 Silent Mutation GCA,GCG A369A NP_001269475.1
NM_001282547.1 1542 Silent Mutation GCA,GCG A364A NP_001269476.1
NM_032017.2 1542 Silent Mutation GCA,GCG A364A NP_114406.1

View Full Product Details