Product Details

SNP ID
rs146588183
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:45341113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAACATCTGGGGAGGAGCTAGT[A/G]GTCCAGGTTCCCGTAGTGGATGTGC
Phenotype
MIM: 604933 MIM: 604746 MIM: 613931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUTYH PubMed Links

Gene Details

Gene
MUTYH
Gene Name
mutY DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048171.1 936 Intron NP_001041636.1
NM_001048172.1 936 Intron NP_001041637.1
NM_001048173.1 936 Intron NP_001041638.1
NM_001048174.1 936 Intron NP_001041639.1
NM_001128425.1 936 Intron NP_001121897.1
NM_001293190.1 936 Intron NP_001280119.1
NM_001293191.1 936 Intron NP_001280120.1
NM_001293192.1 936 Intron NP_001280121.1
NM_001293195.1 936 Intron NP_001280124.1
NM_001293196.1 936 Intron NP_001280125.1
NM_012222.2 936 Intron NP_036354.1
XM_011541497.2 936 Intron XP_011539799.1
XM_011541498.1 936 Intron XP_011539800.1
XM_011541499.1 936 Intron XP_011539801.1
XM_011541500.2 936 Intron XP_011539802.1
XM_011541501.2 936 Intron XP_011539803.1
XM_011541502.2 936 Intron XP_011539804.1
XM_011541503.2 936 Intron XP_011539805.1
XM_011541504.2 936 Intron XP_011539806.1
XM_011541505.2 936 Intron XP_011539807.1
XM_011541506.1 936 Intron XP_011539808.1
XM_011541507.2 936 Intron XP_011539809.2
XM_017001331.1 936 Intron XP_016856820.1
XM_017001332.1 936 Intron XP_016856821.1
XM_017001333.1 936 Intron XP_016856822.1
XM_017001334.1 936 Intron XP_016856823.1
XM_017001335.1 936 Intron XP_016856824.1
XM_017001336.1 936 Intron XP_016856825.1
XM_017001337.1 936 Intron XP_016856826.1
XM_017001338.1 936 Intron XP_016856827.1
XM_017001339.1 936 Intron XP_016856828.1
Gene
TESK2
Gene Name
testis-specific kinase 2
There are no transcripts associated with this gene.

Gene
TOE1
Gene Name
target of EGR1, member 1 (nuclear)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025077.3 936 Silent Mutation GTA,GTG V31V NP_079353.3
XM_005270412.3 936 Silent Mutation GTA,GTG V37V XP_005270469.1
XM_005270413.4 936 UTR 5 XP_005270470.1
XM_011540569.2 936 UTR 5 XP_011538871.1

View Full Product Details