Product Details

SNP ID
rs146590949
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:150152202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATCTAAAAGAACATCTACTGAG[G/T]CATCTAGGGAAGAACTAAGCTGTGC
Phenotype
MIM: 608335 MIM: 610035
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371433 PubMed Links

Gene Details

Gene
LOC105371433
Gene Name
uncharacterized LOC105371433
There are no transcripts associated with this gene.

Gene
PLEKHO1
Gene Name
pleckstrin homology domain containing O1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304722.1 Intron NP_001291651.1
NM_001304723.1 Intron NP_001291652.1
NM_001304724.1 Intron NP_001291653.1
NM_016274.5 Intron NP_057358.2
XM_017001420.1 Intron XP_016856909.1
Gene
VPS45
Gene Name
vacuolar protein sorting 45 homolog
There are no transcripts associated with this gene.

View Full Product Details