Product Details

SNP ID
rs146657303
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:159857836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAAGACCCAGCTCGATAGGGGTA[A/G]TGGTGCCCACTGATCTTGGCCCACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf204 PubMed Links

Gene Details

Gene
C1orf204
Gene Name
chromosome 1 open reading frame 204
There are no transcripts associated with this gene.

Gene
VSIG8
Gene Name
V-set and immunoglobulin domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013661.1 697 Silent Mutation CAC,CAT H187H NP_001013683.1

View Full Product Details