Product Details
- SNP ID
-
rs146678835
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:27978383 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTACCATGGTTACCTGTTTGGC[C/T]GCAATTTCTTCATCTCGTCCATCTC
- Phenotype
-
MIM: 601655
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EYA3
PubMed Links
Gene Details
- Gene
- EYA3
- Gene Name
- EYA transcriptional coactivator and phosphatase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282560.1 |
1974 |
Silent Mutation |
GCA,GCG |
A498A |
NP_001269489.1 |
NM_001282561.1 |
1974 |
Silent Mutation |
GCA,GCG |
A498A |
NP_001269490.1 |
NM_001282562.1 |
1974 |
Silent Mutation |
GCA,GCG |
A491A |
NP_001269491.1 |
NM_001990.3 |
1974 |
Silent Mutation |
GCA,GCG |
A544A |
NP_001981.2 |
XM_006710449.3 |
1974 |
Silent Mutation |
GCA,GCG |
A546A |
XP_006710512.1 |
XM_011540999.2 |
1974 |
Silent Mutation |
GCA,GCG |
A546A |
XP_011539301.1 |
XM_011541001.2 |
1974 |
Silent Mutation |
GCA,GCG |
A544A |
XP_011539303.1 |
XM_011541003.2 |
1974 |
Silent Mutation |
GCA,GCG |
A500A |
XP_011539305.1 |
XM_011541004.2 |
1974 |
Silent Mutation |
GCA,GCG |
A500A |
XP_011539306.1 |
XM_017000652.1 |
1974 |
Silent Mutation |
GCA,GCG |
A546A |
XP_016856141.1 |
XM_017000653.1 |
1974 |
Silent Mutation |
GCA,GCG |
A546A |
XP_016856142.1 |
XM_017000654.1 |
1974 |
Silent Mutation |
GCA,GCG |
A544A |
XP_016856143.1 |
XM_017000655.1 |
1974 |
Silent Mutation |
GCA,GCG |
A544A |
XP_016856144.1 |
XM_017000656.1 |
1974 |
Silent Mutation |
GCA,GCG |
A500A |
XP_016856145.1 |
XM_017000657.1 |
1974 |
Silent Mutation |
GCA,GCG |
A498A |
XP_016856146.1 |
XM_017000658.1 |
1974 |
Silent Mutation |
GCA,GCG |
A491A |
XP_016856147.1 |
XM_017000659.1 |
1974 |
Silent Mutation |
GCA,GCG |
A445A |
XP_016856148.1 |
View Full Product Details