Product Details

SNP ID
rs146684787
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:160190817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGCGGCTGGCACTGCTGTTGCT[A/G]CTGGTGCTAGGGACACCCAAGTCAG
Phenotype
MIM: 607321 MIM: 114250
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP1A4 PubMed Links

Gene Details

Gene
ATP1A4
Gene Name
ATPase Na+/K+ transporting subunit alpha 4
There are no transcripts associated with this gene.

Gene
CASQ1
Gene Name
calsequestrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001231.4 323 Silent Mutation CTA,CTG L22L NP_001222.3

View Full Product Details