Product Details

SNP ID
rs146761460
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:248681849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGCCAGTGTGGACGGCTGCGTA[A/G]GAAAAGCAGCTTAGCCAGGTGGTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR14I1 PubMed Links

Gene Details

Gene
OR14I1
Gene Name
olfactory receptor family 14 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004734.1 591 Silent Mutation TCC,TCT S152S NP_001004734.1
XM_005273132.3 591 Silent Mutation TCC,TCT S152S XP_005273189.1

View Full Product Details