Product Details

SNP ID
rs146773666
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:210237994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCTGTCCATGAATAGATTCTGC[A/G]AGCCCATTGTCTCGGAAGGAGCTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERTAD4 PubMed Links

Gene Details

Gene
SERTAD4
Gene Name
SERTA domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019605.3 413 Missense Mutation AAG,GAG K12E NP_062551.1
XM_005273193.4 413 Missense Mutation AAG,GAG K12E XP_005273250.1
Gene
SERTAD4-AS1
Gene Name
SERTAD4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details