Product Details

SNP ID
rs146790341
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:227816204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCCTCCACGTCTGCGGCGGCT[C/T]CATCCTCAATGAGTACTGGGTGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS38 PubMed Links

Gene Details

Gene
PRSS38
Gene Name
protease, serine 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183062.2 263 Missense Mutation TCC,TTC S88F NP_898885.1
XM_011544175.2 263 Missense Mutation TCC,TTC S88F XP_011542477.1
XM_011544176.2 263 Missense Mutation TCC,TTC S88F XP_011542478.1

View Full Product Details