Product Details

SNP ID
rs146880707
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:40073751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCACTACTTTAGTTAGTTGTCTC[C/G]TTTGGGCCTGGGCACAGTTCTGGCA
Phenotype
MIM: 600722
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CAP1 PubMed Links

Gene Details

Gene
CAP1
Gene Name
adenylate cyclase associated protein 1
There are no transcripts associated with this gene.

Gene
PPT1
Gene Name
palmitoyl-protein thioesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000310.3 1463 UTR 3 NP_000301.1
NM_001142604.1 1463 UTR 3 NP_001136076.1
XM_005271008.1 1463 UTR 3 XP_005271065.1

View Full Product Details