Product Details

SNP ID
rs146952021
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:167809696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACATGTTAGAAATGATTGTCCAC[A/G]GTATTAGCTCTCATCAGGAATTTGT
Phenotype
MIM: 605205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY10 PubMed Links

Gene Details

Gene
ADCY10
Gene Name
adenylate cyclase 10, soluble
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167749.2 6292 Silent Mutation NP_001161221.1
NM_001297772.1 6292 Silent Mutation NP_001284701.1
NM_018417.5 6292 Silent Mutation NP_060887.2
XM_005245330.4 6292 Intron XP_005245387.1
XM_006711449.3 6292 Silent Mutation XP_006711512.1
XM_011509760.2 6292 Silent Mutation XP_011508062.1
XM_011509762.2 6292 Silent Mutation XP_011508064.1
XM_011509763.2 6292 Silent Mutation XP_011508065.1
XM_011509766.2 6292 Intron XP_011508068.1
XM_017001778.1 6292 Intron XP_016857267.1

View Full Product Details