Product Details

SNP ID
rs146966990
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:152827787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGGAGGGGACAGCGGCCAGCA[C/T]TCTGGAGGCTGCTGCTGAAGCGGAC
Phenotype
MIM: 612603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCE1A PubMed Links

Gene Details

Gene
LCE1A
Gene Name
late cornified envelope 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178348.2 315 Silent Mutation CAC,CAT H105H NP_848125.1

View Full Product Details