Product Details

SNP ID
rs146969326
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:1732505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCGCCATGGACAAGGCGACTC[A/G]GTGCAGGGCCTGCGGGACCTGCAGG
Phenotype
MIM: 116951
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK11A PubMed Links

Gene Details

Gene
CDK11A
Gene Name
cyclin dependent kinase 11A
There are no transcripts associated with this gene.

Gene
SLC35E2
Gene Name
solute carrier family 35 member E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199787.1 1174 Intron NP_001186716.1
NM_182838.2 1174 Nonsense Mutation CGA,TGA R251* NP_878258.1

View Full Product Details