Product Details

SNP ID
rs146974230
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:37537783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACTTCTTCCTCCTCCTCCTCAT[C/T]CTCGTCATCTTTCCTGTCTATTTCA
Phenotype
MIM: 608241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNIP1 PubMed Links

Gene Details

Gene
SNIP1
Gene Name
Smad nuclear interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024700.3 1271 Missense Mutation AAT,GAT N386D NP_078976.2

View Full Product Details