Product Details

SNP ID
rs147016490
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:46678476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATGAAGCTAGATACCTAAAGC[C/T]ACCCTTAGCTCCAGGAATGAGTATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EFCAB14 PubMed Links

Gene Details

Gene
EFCAB14
Gene Name
EF-hand calcium binding domain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014774.2 2450 Silent Mutation GTA,GTG V491V NP_055589.1
Gene
EFCAB14-AS1
Gene Name
EFCAB14 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TEX38
Gene Name
testis expressed 38
There are no transcripts associated with this gene.

View Full Product Details