Product Details

SNP ID
rs147056410
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:247672447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTATGGCCAGGGTAATATCAG[C/T]AACATACACCATCACCTCATTGATT
Phenotype
MIM: 611677
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13G1 PubMed Links

Gene Details

Gene
OR13G1
Gene Name
olfactory receptor family 13 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005487.1 595 Missense Mutation ACT,GCT T199A NP_001005487.1

View Full Product Details