Product Details

SNP ID
rs147155903
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:93562391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTGAAATTCAGTCTTGCAGAT[G/T]TCATTCATTTCTTCATCTGGTTGAA
Phenotype
MIM: 604704 MIM: 608848
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCAR3 PubMed Links

Gene Details

Gene
BCAR3
Gene Name
breast cancer anti-estrogen resistance 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261408.1 2219 Missense Mutation GAA,GAC E776D NP_001248337.1
NM_001261409.1 2219 Missense Mutation GAA,GAC E776D NP_001248338.1
NM_001261410.1 2219 Missense Mutation GAA,GAC E685D NP_001248339.1
NM_001308251.1 2219 Missense Mutation GAA,GAC E452D NP_001295180.1
NM_003567.3 2219 Missense Mutation GAA,GAC E776D NP_003558.1
XM_011542251.2 2219 Missense Mutation GAA,GAC E556D XP_011540553.1
XM_017002480.1 2219 Missense Mutation GAA,GAC E776D XP_016857969.1
Gene
FNBP1L
Gene Name
formin binding protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024948.2 2219 Intron NP_001020119.1
NM_001164473.2 2219 Intron NP_001157945.1
NM_017737.4 2219 Intron NP_060207.2
XM_011541625.2 2219 Intron XP_011539927.1
XM_017001533.1 2219 Intron XP_016857022.1

View Full Product Details