Product Details

SNP ID
rs147160883
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:26190724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGAGGACCGTATGGGGTTTGGA[A/G]GGGCAGTAGCTGCACTGAGGGGCCG
Phenotype
MIM: 609121 MIM: 603272
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CATSPER4 PubMed Links

Gene Details

Gene
CATSPER4
Gene Name
cation channel sperm associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198137.1 97 Missense Mutation AGG,GGG R33G NP_937770.1
XM_011541432.2 97 Missense Mutation AGG,GGG R33G XP_011539734.1
XM_011541433.2 97 Missense Mutation AGG,GGG R33G XP_011539735.1
Gene
CNKSR1
Gene Name
connector enhancer of kinase suppressor of Ras 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297647.1 97 Intron NP_001284576.1
NM_001297648.1 97 Intron NP_001284577.1
NM_006314.2 97 Intron NP_006305.2
XM_011540484.2 97 Intron XP_011538786.1
XM_011540485.2 97 Intron XP_011538787.1

View Full Product Details