Product Details
- SNP ID
-
rs147161785
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:21823638 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCGATGTAGACGCTGCCCTTGGC[A/G]TTGACTGCCACGTTGGGACCTGGGG
- Phenotype
-
MIM: 142461
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSPG2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1138469] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSPG2
- Gene Name
- heparan sulfate proteoglycan 2
- Gene
- LDLRAD2
- Gene Name
- low density lipoprotein receptor class A domain containing 2
View Full Product Details