Product Details

SNP ID
rs147211401
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:1402237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCCAGATCCGCTAGGACTTTC[A/C]TGTTGAGCTCCACCTGGCACTGAAA
Phenotype
MIM: 613482 MIM: 611833
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCNL2 PubMed Links

Gene Details

Gene
CCNL2
Gene Name
cyclin L2
There are no transcripts associated with this gene.

Gene
LOC148413
Gene Name
uncharacterized LOC148413
There are no transcripts associated with this gene.

Gene
MRPL20
Gene Name
mitochondrial ribosomal protein L20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318485.1 Intron NP_001305414.1
NM_017971.3 Intron NP_060441.2

View Full Product Details