Product Details

SNP ID
rs147251588
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:153775909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGAGCGGGCAGCGCCGGGAGCC[C/G]GAGATGCAGCGGCCGGAAGCGGCGC
Phenotype
MIM: 611347 MIM: 604193
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
INTS3 PubMed Links

Gene Details

Gene
INTS3
Gene Name
integrator complex subunit 3
There are no transcripts associated with this gene.

Gene
SLC27A3
Gene Name
solute carrier family 27 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317929.1 618 Missense Mutation CGA,GGA R185G NP_001304858.1
NM_024330.1 618 Missense Mutation CGA,GGA R185G NP_077306.1

View Full Product Details