Product Details

SNP ID
rs147270097
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:228408382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTCGAAGTCCAGGAAGATGCCC[A/C]TGTGGCTGGGAGGCTCCATCAGCAT
Phenotype
MIM: 607868 MIM: 606123
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRIM11 PubMed Links

Gene Details

Gene
TRIM11
Gene Name
tripartite motif containing 11
There are no transcripts associated with this gene.

Gene
TRIM17
Gene Name
tripartite motif containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024940.2 2170 Missense Mutation AGG,ATG R418M NP_001020111.1
NM_001134855.1 2170 Intron NP_001128327.1
NM_016102.3 2170 Missense Mutation AGG,ATG R418M NP_057186.1
XM_006711779.2 2170 Missense Mutation AGG,ATG R418M XP_006711842.1
XM_011544209.2 2170 Missense Mutation AGG,ATG R418M XP_011542511.1
XM_011544210.2 2170 Missense Mutation AGG,ATG R418M XP_011542512.1
XM_011544211.2 2170 Missense Mutation AGG,ATG R391M XP_011542513.1
XM_017001418.1 2170 UTR 3 XP_016856907.1
XM_017001419.1 2170 UTR 3 XP_016856908.1

View Full Product Details