Product Details

SNP ID
rs147299257
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:202123254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCAAGGATGGGGGCACCCCAGA[C/G]AGTGGGCAGGAACTGAGGGGCAATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR37L1 PubMed Links

Gene Details

Gene
GPR37L1
Gene Name
G protein-coupled receptor 37 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004767.3 354 Missense Mutation GAC,GAG D97E NP_004758.3
XM_011510158.2 354 Intron XP_011508460.1

View Full Product Details