Product Details

SNP ID
rs147371001
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:1386534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGTGTTTATTCACCATGTCATA[C/T]GCGCATATAGGGAACCTCTATTTAG
Phenotype
MIM: 613482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNL2 PubMed Links

Gene Details

Gene
CCNL2
Gene Name
cyclin L2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039577.3 2176 Intron NP_001034666.1
NM_001320153.1 2176 UTR 3 NP_001307082.1
NM_001320155.1 2176 UTR 3 NP_001307084.1
NM_030937.4 2176 UTR 3 NP_112199.2
XM_011542216.2 2176 UTR 3 XP_011540518.1
XM_011542219.2 2176 UTR 3 XP_011540521.1
XM_011542221.2 2176 UTR 3 XP_011540523.1
XM_017002420.1 2176 UTR 3 XP_016857909.1
XM_017002421.1 2176 UTR 3 XP_016857910.1
XM_017002422.1 2176 UTR 3 XP_016857911.1
XM_017002423.1 2176 Intron XP_016857912.1

View Full Product Details