Product Details

SNP ID
rs147384307
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:17205260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCAGCTGTCCCTGAAGATGCCT[A/G]CCCATGCCGTGTGTGTGGTGGGAGT
Phenotype
MIM: 607934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC400743 PubMed Links

Gene Details

Gene
LOC400743
Gene Name
uncharacterized LOC400743
There are no transcripts associated with this gene.

Gene
PADI1
Gene Name
peptidyl arginine deiminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013358.2 138 Missense Mutation ACC,GCC T15A NP_037490.2
XM_011541307.1 138 Missense Mutation ACC,GCC T15A XP_011539609.1
XM_017001101.1 138 Intron XP_016856590.1
XM_017001102.1 138 Intron XP_016856591.1
XM_017001103.1 138 Missense Mutation ACC,GCC T15A XP_016856592.1

View Full Product Details