Product Details

SNP ID
rs147442172
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:151286498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCCCTGGAGTTCCAGCCCAGGC[C/T]TCTGACCATGGCCTGCCACCGCCAG
Phenotype
MIM: 602758 MIM: 610568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100507670 PubMed Links

Gene Details

Gene
LOC100507670
Gene Name
uncharacterized LOC100507670
There are no transcripts associated with this gene.

Gene
PI4KB
Gene Name
phosphatidylinositol 4-kinase beta
There are no transcripts associated with this gene.

Gene
ZNF687
Gene Name
zinc finger protein 687
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304763.1 509 Silent Mutation GCC,GCT A69A NP_001291692.1
NM_001304764.1 509 Silent Mutation GCC,GCT A69A NP_001291693.1
NM_020832.2 509 Silent Mutation GCC,GCT A69A NP_065883.1
XM_005245366.3 509 Silent Mutation GCC,GCT A78A XP_005245423.1
XM_011509811.2 509 Silent Mutation GCC,GCT A69A XP_011508113.1
XM_011509812.2 509 Silent Mutation GCC,GCT A69A XP_011508114.1
XM_011509813.2 509 Silent Mutation GCC,GCT A69A XP_011508115.1

View Full Product Details