Product Details

SNP ID
rs147520018
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:155175953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAAGTGATCATGTTCCCGTGC[C/G]CAGCCTGCCAAGGTGATGTGGAGCT
Phenotype
MIM: 158340 MIM: 600986
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRTCAP2 PubMed Links

Gene Details

Gene
KRTCAP2
Gene Name
keratinocyte associated protein 2
There are no transcripts associated with this gene.

Gene
MUC1
Gene Name
mucin 1, cell surface associated
There are no transcripts associated with this gene.

Gene
TRIM46
Gene Name
tripartite motif containing 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256599.1 685 Missense Mutation CCA,GCA P108A NP_001243528.1
NM_001256600.1 685 Missense Mutation CCA,GCA P118A NP_001243529.1
NM_001256601.1 685 Missense Mutation CCA,GCA P118A NP_001243530.1
NM_001282378.1 685 Missense Mutation CCA,GCA P5A NP_001269307.1
NM_001282379.1 685 Missense Mutation CCA,GCA P131A NP_001269308.1
NM_025058.4 685 Missense Mutation CCA,GCA P131A NP_079334.3
XM_011510002.1 685 Missense Mutation CCA,GCA P160A XP_011508304.1
XM_017002393.1 685 Missense Mutation CCA,GCA P118A XP_016857882.1

View Full Product Details