Product Details

SNP ID
rs147527561
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:167433055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCCATCGTGCCCCTTGCCCCTC[C/T]GGCGCTGGTTGTTTGGGAGTGAAAT
Phenotype
MIM: 186780 MIM: 164175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD247 PubMed Links

Gene Details

Gene
CD247
Gene Name
CD247 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000734.3 617 Missense Mutation CAG,CGG Q132R NP_000725.1
NM_198053.2 617 Missense Mutation CAG,CGG Q133R NP_932170.1
XM_011510144.2 617 Missense Mutation CAG,CGG Q141R XP_011508446.1
XM_011510145.1 617 Missense Mutation CAG,CGG Q140R XP_011508447.1
XM_017002800.1 617 Missense Mutation CAG,CGG Q164R XP_016858289.1
XM_017002801.1 617 Missense Mutation CAG,CGG Q163R XP_016858290.1
Gene
POU2F1
Gene Name
POU class 2 homeobox 1
There are no transcripts associated with this gene.

View Full Product Details