Product Details

SNP ID
rs147591285
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:1336406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGCTGCTCCCCACCCCACTCGG[A/T]GCCGTGTGATCCGATTCACTGCCAC
Phenotype
MIM: 615467 MIM: 601365 MIM: 605865
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CPTP PubMed Links

Gene Details

Gene
CPTP
Gene Name
ceramide-1-phosphate transfer protein
There are no transcripts associated with this gene.

Gene
DVL1
Gene Name
dishevelled segment polarity protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004421.2 1901 Silent Mutation GCA,GCT A583A NP_004412.2
XM_005244731.3 1901 Silent Mutation GCA,GCT A608A XP_005244788.1
XM_005244732.3 1901 Intron XP_005244789.1
XM_005244733.3 1901 Intron XP_005244790.1
Gene
MIR6808
Gene Name
microRNA 6808
There are no transcripts associated with this gene.

Gene
TAS1R3
Gene Name
taste 1 receptor member 3
There are no transcripts associated with this gene.

View Full Product Details