Product Details

SNP ID
rs147641457
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:205083577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACTGCTGCAGGCCCCCCCTTAG[C/T]GCACAGAGGACAATCCTCACCAACA
Phenotype
MIM: 190197 MIM: 600697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNTN2 PubMed Links

Gene Details

Gene
CNTN2
Gene Name
contactin 2
There are no transcripts associated with this gene.

Gene
RBBP5
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
There are no transcripts associated with this gene.

Gene
TMEM81
Gene Name
transmembrane protein 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203376.1 884 Silent Mutation GCA,GCG A248A NP_976310.1

View Full Product Details