Product Details
- SNP ID
-
rs147642337
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:230911484 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATTACTTACCTGGATCCTCGAGA[A/G]AGTGGCATTAAATTATAAAAGTAAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TTC13
PubMed Links
Gene Details
- Gene
- TTC13
- Gene Name
- tetratricopeptide repeat domain 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001122835.2 |
3344 |
Silent Mutation |
CTC,CTT |
L711L |
NP_001116307.2 |
NM_024525.4 |
3344 |
Silent Mutation |
CTC,CTT |
L765L |
NP_078801.3 |
XM_005273260.1 |
3344 |
Silent Mutation |
CTC,CTT |
L764L |
XP_005273317.1 |
XM_005273261.2 |
3344 |
Silent Mutation |
CTC,CTT |
L734L |
XP_005273318.1 |
XM_005273262.1 |
3344 |
Silent Mutation |
CTC,CTT |
L712L |
XP_005273319.1 |
XM_005273264.2 |
3344 |
Silent Mutation |
CTC,CTT |
L689L |
XP_005273321.1 |
XM_006711814.2 |
3344 |
Silent Mutation |
CTC,CTT |
L636L |
XP_006711877.1 |
XM_006711815.2 |
3344 |
Silent Mutation |
CTC,CTT |
L407L |
XP_006711878.1 |
XM_011544275.2 |
3344 |
Silent Mutation |
CTC,CTT |
L658L |
XP_011542577.1 |
XM_011544276.2 |
3344 |
Silent Mutation |
CTC,CTT |
L468L |
XP_011542578.1 |
XM_017002321.1 |
3344 |
Silent Mutation |
CTC,CTT |
L733L |
XP_016857810.1 |
XM_017002322.1 |
3344 |
Silent Mutation |
CTC,CTT |
L688L |
XP_016857811.1 |
XM_017002323.1 |
3344 |
Silent Mutation |
CTC,CTT |
L636L |
XP_016857812.1 |
XM_017002324.1 |
3344 |
Silent Mutation |
CTC,CTT |
L635L |
XP_016857813.1 |
XM_017002325.1 |
3344 |
Silent Mutation |
CTC,CTT |
L406L |
XP_016857814.1 |
XM_017002326.1 |
3344 |
Silent Mutation |
CTC,CTT |
L406L |
XP_016857815.1 |
View Full Product Details