Product Details
- SNP ID
-
rs147717001
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:151540403 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGAACTGGTGTACCCTGGTGGAC[C/G]TGCACCCAGAGGACCAGGCGGCGGT
- Phenotype
-
MIM: 609473
MIM: 600087
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CGN
PubMed Links
Gene Details
- Gene
- CGN
- Gene Name
- cingulin
There are no transcripts associated with this gene.
- Gene
- MIR554
- Gene Name
- microRNA 554
There are no transcripts associated with this gene.
- Gene
- TUFT1
- Gene Name
- tuftelin 1
View Full Product Details