Product Details
- SNP ID
-
rs147742881
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:1255406 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGTTCTGGACGAGGTGCGTCTCCC[C/T]GTCTGGAACCACGTCTGGCAAGGGG
- Phenotype
-
MIM: 616593
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM132A
PubMed Links
Gene Details
- Gene
- FAM132A
- Gene Name
- family with sequence similarity 132 member A
There are no transcripts associated with this gene.
- Gene
- UBE2J2
- Gene Name
- ubiquitin conjugating enzyme E2 J2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_058167.2 |
639 |
Missense Mutation |
AGG,GGG |
R193G |
NP_477515.2 |
NM_194315.1 |
639 |
Missense Mutation |
AGG,GGG |
R209G |
NP_919296.1 |
NM_194457.1 |
639 |
Missense Mutation |
AGG,GGG |
R141G |
NP_919439.1 |
NM_194458.1 |
639 |
Missense Mutation |
AGG,GGG |
R141G |
NP_919440.1 |
XM_005244718.3 |
639 |
Missense Mutation |
AGG,GGG |
R193G |
XP_005244775.1 |
XM_005244719.4 |
639 |
Missense Mutation |
AGG,GGG |
R193G |
XP_005244776.1 |
XM_006710333.3 |
639 |
Missense Mutation |
AGG,GGG |
R193G |
XP_006710396.1 |
XM_011540613.2 |
639 |
Missense Mutation |
AGG,GGG |
R158G |
XP_011538915.1 |
XM_011540614.2 |
639 |
Missense Mutation |
AGG,GGG |
R193G |
XP_011538916.1 |
XM_011540616.2 |
639 |
Missense Mutation |
AGG,GGG |
R141G |
XP_011538918.1 |
XM_011540617.2 |
639 |
Missense Mutation |
AGG,GGG |
R141G |
XP_011538919.1 |
XM_017000238.1 |
639 |
Missense Mutation |
AGG,GGG |
R141G |
XP_016855727.1 |
XM_017000239.1 |
639 |
Missense Mutation |
AGG,GGG |
R141G |
XP_016855728.1 |
View Full Product Details