Product Details

SNP ID
rs147746078
Assay Type
Functionally Tested
NCBI dbSNP Submissions
13
Location
Chr.1:151729142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGATTAAGGGAATGGAAGTGGAA[A/G]TACAGTAGTTCAGGGGTAAAGATGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RIIAD1 PubMed Links

Gene Details

Gene
RIIAD1
Gene Name
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144956.1 Intron NP_001138428.1

View Full Product Details