Product Details

SNP ID
rs147791058
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:196984091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAAACAATGAGAAAAACATTTC[G/T]TGTGTAGAACGGGGCTGGTCCACTC
Phenotype
MIM: 608593
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CFHR5 PubMed Links

Gene Details

Gene
CFHR5
Gene Name
complement factor H related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030787.3 512 Silent Mutation TCG,TCT S128S NP_110414.1
XM_011510020.2 512 Silent Mutation TCG,TCT S131S XP_011508322.1

View Full Product Details