Product Details

SNP ID
rs147846234
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:19920428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTGTCACACTCGCAGGTCAGC[C/T]GCTGGCAGGTGGTCCTGCCGGCTGG
Phenotype
MIM: 611758
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTUD3 PubMed Links

Gene Details

Gene
OTUD3
Gene Name
OTU deubiquitinase 3
There are no transcripts associated with this gene.

Gene
PLA2G2E
Gene Name
phospholipase A2 group IIE
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014589.2 366 Missense Mutation CAG,CGG Q103R NP_055404.1

View Full Product Details