Product Details

SNP ID
rs147933268
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:156293093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTTCTTGTGGTGCAGCAGCAGA[A/G]CCAAGCCGCCCCCTAGCAGCATGAG
Phenotype
MIM: 610962 MIM: 615531
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLMP PubMed Links

Gene Details

Gene
GLMP
Gene Name
glycosylated lysosomal membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256604.1 875 Missense Mutation GCT,GTT A342V NP_001243533.1
NM_001256605.1 875 Missense Mutation GCT,GTT A305V NP_001243534.1
NM_001256608.1 875 Silent Mutation GGC,GGT G305G NP_001243537.1
NM_001256609.1 875 Missense Mutation GCT,GTT A310V NP_001243538.1
NM_144580.2 875 Missense Mutation GCT,GTT A391V NP_653181.1
XM_011509117.2 875 Missense Mutation GCT,GTT A214V XP_011507419.1
XM_017000149.1 875 UTR 3 XP_016855638.1
Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

Gene
TMEM79
Gene Name
transmembrane protein 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032323.2 875 Intron NP_115699.1
Gene
VHLL
Gene Name
von Hippel-Lindau tumor suppressor like
There are no transcripts associated with this gene.

View Full Product Details