Product Details

SNP ID
rs147939385
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:182451615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAACAGGAGGCAGAGTGCGCAGA[A/G]AGGCAGGAAATGAGACTGGGAGGAA
Phenotype
MIM: 611012
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGSL1 PubMed Links

Gene Details

Gene
RGSL1
Gene Name
regulator of G-protein signaling like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137669.1 Intron NP_001131141.1
XM_005245137.1 Intron XP_005245194.1
XM_011509494.2 Intron XP_011507796.1
XM_011509496.1 Intron XP_011507798.1
XM_011509500.1 Intron XP_011507802.1
XM_011509503.1 Intron XP_011507805.1
XM_011509504.2 Intron XP_011507806.1
XM_017001188.1 Intron XP_016856677.1
XM_017001189.1 Intron XP_016856678.1
XM_017001190.1 Intron XP_016856679.1
XM_017001191.1 Intron XP_016856680.1
XM_017001192.1 Intron XP_016856681.1
XM_017001193.1 Intron XP_016856682.1
XM_017001194.1 Intron XP_016856683.1
XM_017001195.1 Intron XP_016856684.1
XM_017001196.1 Intron XP_016856685.1
XM_017001197.1 Intron XP_016856686.1

View Full Product Details