Product Details

SNP ID
rs147943921
Assay Type
Functionally Tested
NCBI dbSNP Submissions
9
Location
Chr.1:163069343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGTAGCTGGGCTATAAAAGAGAC[C/T]CCTACAGGCTTAGCAGGAAGACGCT
Phenotype
MIM: 602516
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RGS4 PubMed Links

Gene Details

Gene
RGS4
Gene Name
regulator of G-protein signaling 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102445.2 479 Silent Mutation ACC,ACT T50T NP_001095915.1
NM_001113380.1 479 Intron NP_001106851.1
NM_001113381.1 479 UTR 5 NP_001106852.1
NM_005613.5 479 UTR 5 NP_005604.1

View Full Product Details