Product Details
- SNP ID
-
rs147982626
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:47143870 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACCATCTACAGCCTGACCTCTGC[C/T]GGCCGCTGGACACACCGCGCCTGCC
- Phenotype
-
MIM: 615341
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CYP4A22
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10789501,rs911909] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CYP4A22
- Gene Name
- cytochrome P450 family 4 subfamily A member 22
View Full Product Details