Product Details

SNP ID
rs148021104
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:23429857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCATGTGGGGGCCAGCAAGGAGC[C/T]AGTCTTGCAAAAGTTGCACAGAACT
Phenotype
MIM: 616594 MIM: 604128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASAP3 PubMed Links

Gene Details

Gene
ASAP3
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 3
There are no transcripts associated with this gene.

Gene
TCEA3
Gene Name
transcription elongation factor A3
There are no transcripts associated with this gene.

View Full Product Details