Product Details

SNP ID
rs148113334
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:152084996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTGTGGTGCTTGCCTCCCTTT[A/G]TAGTACCAGCCTCTCTCTCTGAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100131107 PubMed Links

Gene Details

Gene
LOC100131107
Gene Name
putative UPF0607 protein ENSP00000383783
There are no transcripts associated with this gene.

Gene
TCHHL1
Gene Name
trichohyalin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008536.1 2751 Nonsense Mutation CAA,TAA Q896* NP_001008536.1

View Full Product Details