Product Details

SNP ID
rs148190221
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:248239609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATGTCCTTCGAGCCGTCATCCA[C/T]ATGGGCTCTGGGGAAAGTCGTCGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 681 Silent Mutation CAC,CAT H227H NP_059974.1

View Full Product Details